Assay Details
Target Gene Details
Entrez Gene ID: | 240549 |
Gene Name: | predicted gene 4952 |
Gene Aliases: |
EG240549 |
Location: |
Chr.19:12599984-12627616 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm4952 | NM_001013762.2 | 1 | 73 | NP_001013784.2 |
NM_001167907.1 | NP_001161379.1 | |||
BC089619.1 | AAH89619.1 |
More Information
Set Membership: |
Intragenic Exonic |