Assay Details
Target Gene Details
Entrez Gene ID: | 14227 |
Gene Name: | FK506 binding protein 2 |
Gene Aliases: |
13kDa, FKBP-2, mFKBP13, mFKBP2 |
Location: |
Chr.19:6977739-6980501 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fkbp2 | NM_001166368.1 | NP_001159840.1 | ||
NM_008020.3 | NP_032046.1 | |||
AK144284.1 | BAE25814.1 | |||
AK165394.1 | BAE38158.1 | |||
BC031824.1 | AAH31824.1 | |||
BC053692.1 | AAH53692.1 | |||
CF949699.1 | ||||
DV655383.1 |