Assay Details
Target Gene Details
Entrez Gene ID: | 12333 |
Gene Name: | calpain 1 |
Gene Aliases: |
Capa-1, Capa1, mu-calpin |
Location: |
Chr.19:5988545-6015825 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Capn1 | NM_001110504.1 | NP_001103974.1 | ||
NM_007600.3 | NP_031626.1 | |||
AF021847.1 | AAB72222.1 | |||
AF084459.1 | AAC33134.1 | |||
AK088547.1 | BAC40416.1 | |||
AK149706.1 | ||||
AK150084.1 | BAE29293.1 | |||
AK160940.1 | BAE36103.1 | |||
AK165258.1 | ||||
AK168063.1 | BAE40039.1 | |||
AK171405.1 | BAE42435.1 | |||
BC026138.1 | AAH26138.1 | |||
BC050276.1 | AAH50276.1 | |||
BY034288.1 |
Target Gene Details
Entrez Gene ID: | 100502985 |
Gene Name: | predicted gene 10814 |
Gene Aliases: |
ENSMUSG00000075230 |
Location: |
Chr.19:6012620-6018459 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm10814 | NR_045783.1 | |||
AK160921.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |