Assay Details
Target Gene Details
Entrez Gene ID: | 58249 |
Gene Name: | fibroblast growth factor (acidic) intracellular binding protein |
Gene Aliases: |
2010004G08Rik, 2010005N21Rik, 3010027N18Rik |
Location: |
Chr.19:5460601-5465052 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fibp | NM_001253832.1 | NP_001240761.1 | ||
NM_021438.4 | NP_067413.1 | |||
AF270700.1 | AAF76201.1 | |||
AK008093.1 | BAB25454.1 | |||
AK008132.1 | ||||
AK012629.1 | BAB28367.1 | |||
AK019399.1 | ||||
AK028029.1 | BAC25708.1 | |||
AK090037.1 | BAC41059.1 | |||
BC031716.1 | AAH31716.1 | |||
BC049284.1 | AAH49284.1 |