Assay Details
Target Gene Details
Entrez Gene ID: | 11632 |
Gene Name: | aryl-hydrocarbon receptor-interacting protein |
Gene Aliases: |
AA408703, AW476050, Ara9, D19Bwg1412e, Fkbp16, Xap2 |
Location: |
Chr.19:4113756-4127236 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aip | NM_001276284.1 | NP_001263213.1 | ||
NM_016666.3 | NP_057875.1 | |||
AK011609.1 | ||||
AK146390.1 | BAE27133.1 | |||
BC075614.1 | AAH75614.1 | |||
BQ175443.1 | ||||
U85489.1 | AAB59009.1 |
More Information
Set Membership: |
Intragenic Non-exonic |