Assay Details
Target Gene Details
Entrez Gene ID: | 56248 |
Gene Name: | adenylate kinase 3 |
Gene Aliases: |
AA407498, AI506714, AK-3, Ak3l, Ak3l1, Akl3l |
Location: |
Chr.19:28995380-29048672 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ak3 |