Assay Details
Target Gene Details
Entrez Gene ID: | 104174 |
Gene Name: | glycine decarboxylase |
Gene Aliases: |
D030049L12Rik, D19Wsu57e |
Location: |
Chr.19:30098441-30175441 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gldc | NM_138595.2 | 18 | 2329 | NP_613061.1 |
AK080789.1 | 18 | 2301 | BAC38022.1 | |
AK080937.1 | 18 | 2319 | ||
AK139929.1 | 18 | 2322 | ||
BC017135.1 | 18 | 2306 | AAH17135.1 |
More Information
Set Membership: |
Intragenic Exonic |