Assay Details
Target Gene Details
Entrez Gene ID: | 67160 |
Gene Name: | eukaryotic translation elongation factor 1 gamma |
Gene Aliases: |
2610301D06Rik, AA407312, EF1G |
Location: |
Chr.19:8967041-8978180 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eef1g | NM_026007.4 | NP_080283.3 | ||
AF321126.1 | AAL23895.1 | |||
AK007869.1 | BAB25320.1 | |||
AK007991.1 | ||||
AK011951.1 | ||||
AK011973.1 | BAB27948.1 | |||
AK012431.1 | ||||
AK014277.1 | BAB29238.3 | |||
AK050636.1 | BAC34356.1 | |||
AK081859.1 | BAC38351.1 | |||
AK145519.1 | BAE26482.1 | |||
AK145565.1 | BAE26514.1 | |||
AK147980.1 | BAE28265.1 | |||
AK159644.1 | ||||
AK159768.1 | ||||
AK166765.1 | BAE39003.1 | |||
AK167831.1 | BAE39853.1 | |||
AK168749.1 | BAE40589.1 | |||
AK183747.1 | ||||
AK192634.1 | ||||
BC023495.1 | AAH23495.1 | |||
BC083071.1 | AAH83071.1 | |||
BC099413.1 | AAH99413.1 | |||
BC117547.1 | ||||
BC118522.1 |