Assay Details
Target Gene Details
Entrez Gene ID: | 68693 |
Gene Name: | heterogeneous nuclear ribonucleoprotein U-like 2 |
Gene Aliases: |
1110031M08Rik, AI465155, Hnrpul2 |
Location: |
Chr.19:8819401-8834142 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hnrnpul2 | NM_001081196.1 | NP_001074665.1 | ||
DQ000354.1 |