Assay Details
Target Gene Details
Entrez Gene ID: | 319924 |
Gene Name: | amyloid beta (A4) precursor protein binding, family A, member 1 |
Gene Aliases: |
6430513E09Rik, Lin-10, Mint, Mint1, Urop11, X11, X11alpha |
Location: |
Chr.19:23758876-23949598 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apba1 | NM_177034.3 | 8 | 2031 | NP_796008.2 |
AK032261.1 | 7 | 795 | BAC27784.1 | |
AK147583.1 | 8 | 2021 | BAE28008.1 | |
AK147623.1 | 8 | 1983 | BAE28031.1 | |
BC141181.1 | 8 | 1849 |
More Information
Set Membership: |
Intragenic Exonic |