Assay Details
Target Gene Details
Entrez Gene ID: | 269016 |
Gene Name: | SH3 domain containing ring finger 2 |
Gene Aliases: |
2310046K19, 9130023G24Rik, AI452083, Ppp1r39, RNF158 |
Location: |
Chr.18:42051743-42158695 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sh3rf2 | NM_001146299.1 | NP_001139771.1 | ||
NM_172966.3 | NP_766554.2 | |||
AK028153.1 | BAC25780.1 | |||
AK029168.1 | BAC26334.1 | |||
AK033601.1 | BAC28382.1 | |||
BC137955.1 | ||||
BC145060.1 |