Assay Details
Target Gene Details
Entrez Gene ID: | 381126 |
Gene Name: | GRB2 associated, regulator of MAPK1 |
Gene Aliases: |
C86169, Fam59a, Gm944, mKIAA4238 |
Location: |
Chr.18:21127342-21300139 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Garem | NM_001033445.2 | NP_001028617.2 | ||
AK148318.1 | BAE28477.1 | |||
AK220564.1 | BAD90542.1 |