Assay Details
Target Gene Details
Entrez Gene ID: | 70417 |
Gene Name: | multiple EGF-like-domains 10 |
Gene Aliases: |
3000002B06Rik, Gm331 |
Location: |
Chr.18:57133090-57297467 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Megf10 | NM_001001979.2 | 10 | 1875 | NP_001001979.1 |
AK147238.1 | 10 | 1875 | BAE27788.1 | |
AK148084.1 | 10 | 1438 | BAE28336.1 | |
AK166316.1 | 10 | 1472 | BAE38699.1 | |
BC075647.1 | 10 | 1490 | AAH75647.1 |
More Information
Set Membership: |
Intragenic Exonic |