Assay Details
Target Gene Details
Entrez Gene ID: | 269003 |
Gene Name: | Sin3A associated protein |
Gene Aliases: |
2610304F09Rik, 6720406D06 |
Location: |
Chr.18:31634327-31723068 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sap130 | NM_172965.2 | 5 | 669 | NP_766553.1 |
AK034401.1 | 5 | 673 | ||
AK053675.1 | 5 | 670 | BAC35469.1 | |
AK084078.1 | 5 | 670 | BAC39113.1 | |
BC063075.1 | 2 | 193 | AAH63075.1 | |
BC066030.1 | 5 | 681 | AAH66030.1 |
More Information
Set Membership: |
Intragenic Exonic |