Assay Details
Target Gene Details
Entrez Gene ID: | 225326 |
Gene Name: | phosphoinositide-3-kinase, class 3 |
Gene Aliases: |
5330434F23Rik, Vps34 |
Location: |
Chr.18:30272094-30348126 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pik3c3 | NM_181414.5 | NP_852079.2 | ||
AK030578.1 | ||||
AK044997.1 | ||||
BC024675.1 | AAH24675.1 | |||
BC057678.1 | AAH57678.1 |
More Information
Set Membership: |
Intragenic Non-exonic |