Assay Details
Target Gene Details
Entrez Gene ID: | 18145 |
Gene Name: | Niemann-Pick type C1 |
Gene Aliases: |
A430089E03Rik, C85354, D18Ertd139e, D18Ertd723e, lcsd, nmf164, spm |
Location: |
Chr.18:12189694-12236386 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Npc1 | NM_008720.2 | 9 | 1589 | NP_032746.2 |
AF003348.1 | 9 | 1566 | AAB63372.1 | |
AK154282.1 | 9 | 1509 | BAE32486.1 | |
AK155379.1 | 9 | 1509 | BAE33230.1 | |
BC052437.1 | 9 | 1589 | AAH52437.1 | |
BC054539.1 | 9 | 1589 | AAH54539.1 |