Assay Details
Target Gene Details
Entrez Gene ID: | 19348 |
Gene Name: | kinesin family member 20A |
Gene Aliases: |
AA415432, Rab6kifl |
Location: |
Chr.18:34624624-34633277 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kif20a | NM_001166406.1 | 5 | 698 | NP_001159878.1 |
NM_001166407.1 | 5 | 696 | NP_001159879.1 | |
NM_009004.4 | 5 | 592 | NP_033030.1 | |
AK051201.1 | 5 | 510 | ||
AK051995.1 | 5 | 547 | ||
AK053280.1 | 5 | 510 | ||
AK083412.1 | 5 | 552 | BAC38906.1 | |
AK083697.1 | 5 | 512 | ||
AK084732.1 | 5 | 534 | BAC39267.1 | |
AK142764.1 | 5 | 522 | ||
AK144844.1 | 5 | 510 | BAE26095.1 | |
BC060608.1 | 5 | 698 | AAH60608.1 | |
Y09632.1 | 5 | 747 | CAA70845.1 |
More Information
Set Membership: |
Intragenic Exonic |