Assay Details
Target Gene Details
Entrez Gene ID: | 225467 |
Gene Name: | protein geranylgeranyltransferase type I, beta subunit |
Gene Aliases: |
2010207C17Rik, 2610100E13, AI451237, AI551093, BGG1, GGT1 |
Location: |
Chr.18:46235474-46281089 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pggt1b | NM_172627.3 | NP_766215.1 | ||
AK028188.1 | ||||
AK081731.1 | BAC38314.1 | |||
BC048404.1 | ||||
BC141021.1 |
More Information
Set Membership: |
Intragenic Non-exonic |