Assay Details
Target Gene Details
Entrez Gene ID: | 19348 |
Gene Name: | kinesin family member 20A |
Gene Aliases: |
AA415432, Rab6kifl |
Location: |
Chr.18:34624624-34633277 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kif20a | NM_001166406.1 | NP_001159878.1 | ||
NM_001166407.1 | NP_001159879.1 | |||
NM_009004.4 | NP_033030.1 | |||
AK051201.1 | ||||
AK051995.1 | ||||
AK053280.1 | ||||
AK083412.1 | BAC38906.1 | |||
AK083697.1 | ||||
AK084732.1 | BAC39267.1 | |||
AK142764.1 | ||||
AK144844.1 | BAE26095.1 | |||
BC060608.1 | AAH60608.1 | |||
BY012551.1 | ||||
Y09632.1 | CAA70845.1 |
More Information
Set Membership: |
Intragenic Exonic |