Assay Details
Target Gene Details
Entrez Gene ID: | 15200 |
Gene Name: | heparin-binding EGF-like growth factor |
Gene Aliases: |
AW047313, Dtr, Dts, Hegfl |
Location: |
Chr.18:36504927-36515805 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hbegf | NM_010415.2 | NP_034545.1 | ||
AK145548.1 | BAE26501.1 | |||
AK160575.1 | BAE35883.1 | |||
AK164309.1 | BAE37729.1 | |||
BC089607.1 | AAH89607.1 | |||
L07264.1 | AAA37542.1 |
More Information
Set Membership: |
Intragenic Non-exonic |