Assay Details
Target Gene Details
Entrez Gene ID: | 225288 |
Gene Name: | formin homology 2 domain containing 3 |
Gene Aliases: |
A930009H06Rik, FHOS2, mKIAA1695 |
Location: |
Chr.18:24708456-25133507 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fhod3 | NM_001289654.1 | 24 | 5111 | NP_001276583.1 |
NM_001289655.1 | 25 | 5468 | NP_001276584.1 | |
NM_175276.4 | 26 | 5564 | NP_780485.2 | |
AB078608.1 | 26 | 4742 | BAC98302.1 | |
AB078609.1 | 24 | 4289 | BAC98303.1 | |
AB100088.1 | 25 | 4646 | BAC98348.1 | |
AK220413.1 | 22 | 3728 | BAD90459.1 | |
BC060607.1 | 11 | 2520 | AAH60607.1 | |
BC138046.1 | 24 | 4211 |
Target Gene Details
Entrez Gene ID: | 66648 |
Gene Name: | tubulin polyglutamylase complex subunit 2 |
Gene Aliases: |
5730437P09Rik, 5730494M16Rik, AI666318, Pgs2 |
Location: |
Chr.18:25127223-25169007 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tpgs2 | NM_001142698.1 | NP_001136170.1 | ||
AK141632.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |