Assay Details
Target Gene Details
Entrez Gene ID: | 24068 |
Gene Name: | steroid receptor RNA activator 1 |
Gene Aliases: |
AA959952, Sra, Srap, Straa1, Strra1 |
Location: |
Chr.18:36667187-36670311 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sra1 | NM_001164406.1 | NP_001157878.1 | ||
NM_025291.3 | 4 | 438 | NP_079567.2 | |
NR_028355.1 | 3 | 556 | ||
AF092039.1 | 3 | 342 | ||
AK007302.1 | 4 | 424 | BAB24943.1 | |
AK010375.1 | 4 | 424 | BAB26893.1 | |
AK154232.1 | 4 | 408 | BAE32451.1 | |
AK180031.1 | ||||
BC026480.1 | 2 | 217 | AAH26480.1 | |
BC048362.1 | 4 | 410 | AAH48362.1 | |
BQ748094.1 | 2 | 106 | ||
BX516162.1 |