Assay Details
Target Gene Details
Entrez Gene ID: | 18596 |
Gene Name: | platelet derived growth factor receptor, beta polypeptide |
Gene Aliases: |
AI528809, CD140b, PDGFR-1, Pdgfr |
Location: |
Chr.18:61045150-61085068 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdgfrb | NM_001146268.1 | NP_001139740.1 | ||
NM_008809.2 | NP_032835.2 | |||
AK134785.1 | BAE22283.1 | |||
AK143247.1 | ||||
AK158145.1 | BAE34383.1 | |||
BC055311.1 | AAH55311.1 | |||
DQ979390.1 | ||||
X04367.1 | CAA27882.1 |
More Information
Set Membership: |
Intragenic Non-exonic |