Assay Details
Target Gene Details
Entrez Gene ID: | 225363 |
Gene Name: | eukaryotic translation termination factor 1 |
Gene Aliases: |
AI463371, D6Ertd109e, ERF, ERF1, SUP45L1, TB3-1 |
Location: |
Chr.18:34902785-34932003 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Etf1 | NM_144866.3 | 3 | 319 | NP_659115.3 |
AK049607.1 | 3 | 270 | BAC33839.1 | |
AK077559.1 | 3 | 267 | ||
AK139933.1 | 3 | 280 | ||
AK151028.1 | 3 | 219 | ||
AK151710.1 | 3 | 219 | ||
AK152428.1 | 3 | 219 | BAE31210.1 | |
AK152952.1 | 3 | 219 | BAE31619.1 | |
AK164020.1 | 3 | 270 | BAE37589.1 | |
AK166740.1 | 3 | 288 | BAE38983.1 | |
AK169345.1 | 3 | 292 | BAE41096.1 | |
AK170221.1 | 3 | 262 | BAE41644.1 | |
BC013717.1 | 3 | 246 | AAH13717.1 | |
BY238984.1 | 3 | 319 |
More Information
Set Membership: |
Intragenic Exonic |