Assay Details
Target Gene Details
Entrez Gene ID: | 14164 |
Gene Name: | fibroblast growth factor 1 |
Gene Aliases: |
Dffrx, Fam, Fgf-1, Fgfa |
Location: |
Chr.18:38838672-38929470 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fgf1 | NM_010197.3 | 2 | 259 | NP_034327.1 |
AF012926.1 | 2 | 302 | AAB94020.1 | |
AK035330.1 | 2 | 260 | BAC29035.1 | |
AK142163.1 | 2 | 246 | ||
BC037601.1 | 2 | 278 | AAH37601.1 | |
M30641.1 | 1 | 72 | AAA37618.1 | |
U67610.1 | 2 | 225 | AAC52907.1 |
More Information
Set Membership: |
Intragenic Exonic |