Assay Details
Target Gene Details
Entrez Gene ID: | 77805 |
Gene Name: | establishment of cohesion 1 homolog 1 (S. cerevisiae) |
Gene Aliases: |
A930014I12Rik |
Location: |
Chr.18:10566507-10610685 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Esco1 | NM_001081222.1 | 2 | 388 | NP_001074691.1 |
AK020858.1 | 2 | 189 | ||
AK040576.1 | 2 | 146 | ||
AK045656.1 | 2 | 110 | BAC32447.1 | |
AK049589.1 | 2 | 146 | BAC33830.1 | |
AK139549.1 | 2 | 112 | ||
AK145333.1 | 2 | 389 | BAE26373.1 | |
AK145870.1 | 2 | 166 | BAE26712.1 | |
BC151069.1 | 2 | 344 | ||
BC151077.1 | 2 | 344 |
More Information
Set Membership: |
Intragenic Exonic |