Assay Details
Target Gene Details
Entrez Gene ID: | 19123 |
Gene Name: | protein C |
Gene Aliases: |
PC |
Location: |
Chr.18:32123126-32139570 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Proc | NM_001042767.2 | NP_001036232.1 | ||
NM_001042768.2 | NP_001036233.2 | |||
NM_008934.3 | NP_032960.3 | |||
NR_045173.1 | ||||
AF318182.1 | AAK07918.1 | |||
AI286922.1 | ||||
BC013896.1 | AAH13896.1 | |||
CA468822.1 | ||||
CB599669.1 | ||||
CF550816.1 | ||||
D10445.1 | BAA01235.1 | |||
DV662175.1 |
More Information
Set Membership: |
Intragenic Non-exonic |