Assay Details
Target Gene Details
Entrez Gene ID: | 19123 |
Gene Name: | protein C |
Gene Aliases: |
PC |
Location: |
Chr.18:32123126-32139570 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Proc | NM_001042767.2 | 3 | 199 | NP_001036232.1 |
NM_001042768.2 | 4 | 279 | NP_001036233.2 | |
NM_008934.3 | 3 | 211 | NP_032960.3 | |
NR_045173.1 | 3 | 215 | ||
AF318182.1 | 3 | 188 | AAK07918.1 | |
AI286922.1 | 3 | 199 | ||
BC013896.1 | 3 | 216 | AAH13896.1 | |
BY082597.1 | 3 | 185 | ||
BY086829.1 | 3 | 212 | ||
BY788141.1 | 4 | 256 | ||
CA468822.1 | 3 | 223 | ||
CB599669.1 | 3 | 202 | ||
D10445.1 | 2 | 127 | BAA01235.1 | |
DV662175.1 | 2 | 113 |
More Information
Set Membership: |
Intragenic Exonic |