Assay Details
Target Gene Details
Entrez Gene ID: | 225280 |
Gene Name: | INO80 complex subunit C |
Gene Aliases: |
D030070L09Rik |
Location: |
Chr.18:24104761-24121819 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ino80c | NM_172625.2 | NP_766213.1 | ||
AK077290.1 | BAC36733.1 | |||
AK083718.1 | BAC39004.1 | |||
BC060961.1 | AAH60961.1 |