Assay Details
Target Gene Details
Entrez Gene ID: | 13506 |
Gene Name: | desmocollin 2 |
Gene Aliases: |
AW228162, Dsc2a, Dsc2b |
Location: |
Chr.18:20030633-20059552 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dsc2 | NM_013505.3 | 3 | 514 | NP_038533.1 |
AK028604.1 | 3 | 519 | ||
AK028636.1 | 3 | 515 | BAC26041.1 | |
AK030333.1 | 3 | 515 | BAC26907.1 | |
AK048547.1 | 3 | 515 | BAC33371.1 | |
AK172168.1 | 3 | 349 | BAE42862.1 | |
BC057867.1 | 3 | 294 | AAH57867.1 | |
L33779.1 | 3 | 313 | AAA79176.1 |
More Information
Set Membership: |
Intragenic Exonic |