Assay Details
Target Gene Details
Entrez Gene ID: | 212307 |
Gene Name: | microtubule-associated protein, RP/EB family, member 2 |
Gene Aliases: |
AI314113, C820009F03Rik, D18Abb1e, EB1, EB2, RP1 |
Location: |
Chr.18:23752333-23893861 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mapre2 | NM_001162941.1 | 5 | 529 | NP_001156413.1 |
NM_001162942.1 | 5 | 472 | NP_001156414.1 | |
NM_153058.4 | 4 | 580 | NP_694698.3 | |
AI325879.1 | 5 | 466 | ||
AK028823.1 | 5 | 530 | BAC26138.1 | |
AK038562.1 | 4 | 567 | BAC30045.1 | |
AK040767.1 | 4 | 555 | BAC30700.1 | |
AK045493.1 | 4 | 463 | BAC32393.1 | |
AK045904.1 | 4 | 555 | BAE43326.1 | |
AK046095.1 | 4 | 522 | ||
AK046221.1 | 4 | 547 | BAC32643.1 | |
AK148954.1 | 4 | 555 | BAE28700.1 | |
AK168834.1 | 6 | 414 | BAE40658.1 | |
AK192344.1 | 2 | 164 | ||
BC025804.1 | 4 | 527 | AAH25804.1 | |
BC027056.1 | 4 | 546 | AAH27056.1 | |
BC028987.1 | 4 | 502 | AAH28987.1 | |
BC035254.1 | 4 | 532 | AAH35254.1 |
More Information
Set Membership: |
Intragenic Exonic |