Assay Details
Target Gene Details
Entrez Gene ID: | 22240 |
Gene Name: | dihydropyrimidinase-like 3 |
Gene Aliases: |
CRMP-4, DRP-3, TUC4, ULIP-1, Ulip, Ulip1 |
Location: |
Chr.18:43320979-43438286 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dpysl3 | NM_001136086.2 | NP_001129558.1 | ||
NM_001291455.1 | NP_001278384.1 | |||
NM_009468.5 | NP_033494.1 | |||
AK077000.1 | ||||
AK133040.1 | BAE21480.1 | |||
AK149024.1 | ||||
AK161509.1 | BAE36433.1 | |||
AK171653.1 | BAE42590.1 | |||
BC023003.1 | AAH23003.1 | |||
X87817.1 | CAA61082.1 |
More Information
Set Membership: |
Intragenic Non-exonic |