Assay Details
Target Gene Details
Entrez Gene ID: | 22240 |
Gene Name: | dihydropyrimidinase-like 3 |
Gene Aliases: |
CRMP-4, DRP-3, TUC4, ULIP-1, Ulip, Ulip1 |
Location: |
Chr.18:43320979-43438286 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dpysl3 | NM_001136086.2 | 4 | 561 | NP_001129558.1 |
NM_001291455.1 | 4 | 811 | NP_001278384.1 | |
NM_009468.5 | 4 | 742 | NP_033494.1 | |
AK077000.1 | 4 | 552 | ||
AK133040.1 | 4 | 561 | BAE21480.1 | |
AK149024.1 | 4 | 580 | ||
AK161509.1 | 4 | 483 | BAE36433.1 | |
BC023003.1 | 4 | 426 | AAH23003.1 | |
X87817.1 | 4 | 702 | CAA61082.1 |
More Information
Set Membership: |
Intragenic Exonic |