Assay Details
Target Gene Details
Entrez Gene ID: | 20975 |
Gene Name: | synaptojanin 2 |
Gene Aliases: |
AI481647, SJ2, mKIAA0348 |
Location: |
Chr.17:5941280-6045553 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Synj2 | NM_001113351.1 | 11 | 1701 | NP_001106822.1 |
NM_001113352.2 | 12 | 1790 | NP_001106823.1 | |
NM_001113353.2 | 12 | 1790 | NP_001106824.1 | |
NM_001290698.1 | 6 | 818 | NP_001277627.1 | |
NM_011523.2 | 11 | 1701 | NP_035653.2 | |
AF041857.1 | 12 | 1791 | AAC40141.1 | |
AF041858.1 | 1 | 53 | AAC40142.1 | |
AF041859.1 | 5 | 582 | AAC40143.1 | |
AF041862.1 | 11 | 1646 | AAC40146.1 | |
AK014584.1 | 8 | 945 | ||
AK019694.1 | 6 | 819 | BAB31837.1 | |
AK029755.1 | 9 | 1208 | ||
AK038038.1 | 9 | 1114 | ||
AK129122.1 | 10 | 1218 | BAC97932.1 | |
AK142919.1 | 11 | 1422 | ||
AK148053.1 | 12 | 1529 | ||
AK161467.1 | 6 | 802 | BAE36412.1 | |
AK163512.1 | 10 | 1431 | ||
BC058749.1 | 12 | 1667 | AAH58749.1 | |
BC060214.1 | 11 | 1547 | AAH60214.1 |
More Information
Set Membership: |
Intragenic Exonic |