Assay Details
Target Gene Details
Entrez Gene ID: | 81877 |
Gene Name: | tenascin XB |
Gene Aliases: |
Tn-mhc, Tnx |
Location: |
Chr.17:34659788-34795460 on Build GRCm38 |
Assay Gene Location: | Within Exon 33 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tnxb | NM_031176.2 | 33 | 9413 | NP_112453.2 |
AB235155.1 | BAE44473.1 | |||
AK136535.1 | 21 | 4510 | ||
AK171696.1 | 20 | 4148 | BAE42616.1 | |
X73959.1 | 15 | 2615 | CAA52147.1 |
More Information
Set Membership: |
Intragenic Exonic |