Assay Details
Target Gene Details
Entrez Gene ID: | 11789 |
Gene Name: | adenomatosis polyposis coli |
Gene Aliases: |
AI047805, AU020952, AW124434, CC1, Min, mAPC |
Location: |
Chr.18:34220307-34322190 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apc | NM_007462.3 | NP_031488.2 | ||
AK046280.1 | ||||
AK080907.1 | BAC38073.1 | |||
BC141141.1 | ||||
M88127.1 | AAB59632.1 |