Assay Details
Target Gene Details
Entrez Gene ID: | 21354 |
Gene Name: | transporter 1, ATP-binding cassette, sub-family B (MDR/TAP) |
Gene Aliases: |
ABC17, APT1, Abcb2, Ham-1, Ham1, MTP1, PSF1, RING4, TAP, Tap-1, Y3 |
Location: |
Chr.17:34187556-34197225 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tap1 | NM_001161730.1 | 8 | 1679 | NP_001155202.1 |
NM_013683.2 | 7 | 1763 | NP_038711.2 | |
AK040919.1 | 5 | 2726 | ||
AK151481.1 | 7 | 1518 | ||
AK152953.1 | 7 | 1463 | BAE31620.1 | |
AK153890.1 | 7 | 1767 | BAE32237.1 | |
AK162731.1 | 7 | 1563 | BAE37041.1 | |
AK166046.1 | 7 | 1764 | ||
AK167485.1 | 7 | 1765 | ||
AK171363.1 | 8 | 1434 | BAE42411.1 | |
BC013802.1 | 7 | 1174 | AAH13802.1 | |
BC024897.1 | 7 | 1286 | AAH24897.1 | |
KJ650227.1 | 7 | 1439 | ||
M55637.1 | 7 | 1155 | AAA39570.1 | |
U60018.1 | 7 | 1439 | AAB41962.1 | |
U60019.1 | 7 | 1439 | AAB41963.1 | |
U60020.1 | 7 | 1439 | AAB41964.1 | |
U60021.1 | 7 | 1439 | AAB41965.1 | |
U60022.1 | 7 | 1439 | AAB41966.1 | |
U60023.1 | 7 | 1439 | AAB41967.1 | |
X59615.1 | 7 | 998 | CAA42178.1 |
More Information
Set Membership: |
Intragenic Exonic |