Assay Details
Target Gene Details
Entrez Gene ID: | 18140 |
Gene Name: | ubiquitin-like, containing PHD and RING finger domains, 1 |
Gene Aliases: |
AL022808, ICBP90, Np95, RNF106 |
Location: |
Chr.17:56303337-56323486 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Uhrf1 | NM_001111079.1 | NP_001104549.1 | ||
NM_010931.3 | NP_035061.3 | |||
AF274046.1 | AAK55743.1 | |||
AK145543.1 | BAE26496.1 | |||
AK146951.1 | BAE27560.1 | |||
AK147046.1 | BAE27632.1 | |||
BC022167.1 | AAH22167.1 | |||
BY140812.1 | ||||
D87908.1 | BAA74579.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |