Assay Details
Target Gene Details
Entrez Gene ID: | 269033 |
Gene Name: | RIKEN cDNA 4930503L19 gene |
Gene Aliases: |
9930107F24, BB148262, Lars, Las2 |
Location: |
Chr.18:70452210-70472504 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4930503L19Rik | NM_172967.2 | NP_766555.1 | ||
AK037066.1 | BAC29691.1 | |||
AK080780.1 | 1 | 90 | BAC38019.1 | |
AK087788.1 | BAC40002.1 | |||
AK148410.1 | 1 | 85 | BAE28537.1 | |
AK160714.1 |
Target Gene Details
Entrez Gene ID: | 170461 |
Gene Name: | StAR-related lipid transfer (START) domain containing 6 |
Gene Aliases: |
1700011K09Rik, 4833424I06Rik, 4933429L05Rik, AI644424 |
Location: |
Chr.18:70472454-70505254 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Stard6 | NM_029019.4 | NP_083295.1 |