Assay Details
Target Gene Details
Entrez Gene ID: | 224826 |
Gene Name: | ubiquitin protein ligase E3 component n-recognin 2 |
Gene Aliases: |
9930021A08Rik, AI462103, AW540746, E130209G04Rik, mKIAA0349 |
Location: |
Chr.17:46928291-47010775 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ubr2 | NM_001177374.1 | 23 | 2830 | NP_001170845.1 |
NM_146078.3 | 23 | 2830 | NP_666190.2 | |
AK083320.1 | 23 | 2969 | BAC38864.1 | |
AK122254.2 | 21 | 3659 | BAC65536.3 | |
AK143202.1 | 23 | 2830 | BAE25301.1 | |
AK147445.1 | 23 | 2819 | BAE27917.1 | |
AY061885.1 | 23 | 2517 | AAL32102.1 | |
AY280958.1 | 23 | 2545 | AAQ17202.1 | |
BC031403.1 | 8 | 624 | AAH31403.1 | |
BC075642.1 | 24 | 2644 | AAH75642.1 |
More Information
Set Membership: |
Intragenic Exonic |