Assay Details
Target Gene Details
Entrez Gene ID: | 74094 |
Gene Name: | tight junction associated protein 1 |
Gene Aliases: |
0610041D19Rik, AI415281, AW121008, Pilt, Tjp4 |
Location: |
Chr.17:46257851-46283065 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tjap1 | NM_001252473.1 | NP_001239402.1 | ||
NM_001252474.1 | NP_001239403.1 | |||
NM_001252475.1 | NP_001239404.1 | |||
NM_028751.3 | NP_083027.1 | |||
AB712252.1 | ||||
AK002882.2 | BAB22429.1 | |||
AK164925.1 | ||||
AK169889.1 | BAE41438.1 | |||
AK177408.1 | ||||
BC023316.1 | AAH23316.1 |
More Information
Set Membership: |
Intragenic Non-exonic |