Assay Details
Target Gene Details
Entrez Gene ID: | 195359 |
Gene Name: | tripartite motif-containing 40 |
Gene Aliases: |
Gm319, Gm35, Gm865 |
Location: |
Chr.17:36881598-36896743 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trim40 | NM_001033235.3 | NP_001028407.1 | ||
NM_001172168.1 | NP_001165639.1 | |||
AK136498.1 | BAE23012.1 | |||
BC147336.1 | ||||
BC147337.1 | ||||
BC172037.1 |
More Information
Set Membership: |
Intragenic Non-exonic |