Assay Details
Target Gene Details
Entrez Gene ID: | 20975 |
Gene Name: | synaptojanin 2 |
Gene Aliases: |
AI481647, SJ2, mKIAA0348 |
Location: |
Chr.17:5941280-6045553 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Synj2 | NM_001113351.1 | 3 | 677 | NP_001106822.1 |
NM_001113352.2 | 4 | 766 | NP_001106823.1 | |
NM_001113353.2 | 4 | 766 | NP_001106824.1 | |
NM_011523.2 | 3 | 677 | NP_035653.2 | |
AF041857.1 | 4 | 767 | AAC40141.1 | |
AF041862.1 | 3 | 622 | AAC40146.1 | |
AK029755.1 | 1 | 184 | ||
AK129122.1 | 2 | 194 | BAC97932.1 | |
AK142919.1 | 3 | 398 | ||
AK148053.1 | 4 | 505 | ||
AK163512.1 | 2 | 407 | ||
BC058749.1 | 4 | 643 | AAH58749.1 | |
BC060214.1 | 3 | 523 | AAH60214.1 |
More Information
Set Membership: |
Intragenic Exonic |