Assay Details
Target Gene Details
Entrez Gene ID: | 76482 |
Gene Name: | RIKEN cDNA 3110002H16 gene |
Gene Aliases: |
2400010D15Rik, Mic-1, Mic1 |
Location: |
Chr.18:12168691-12198497 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
3110002H16Rik | NM_029623.2 | 20 | 2003 | NP_083899.1 |
AF003352.1 | 8 | 889 | AAB63375.1 | |
AK013979.1 | 14 | 1389 | ||
AK144623.1 | 20 | 2004 | BAE25975.1 | |
AK154240.1 | 20 | 1998 | BAE32456.1 | |
AK159691.1 | 20 | 1998 | BAE35292.1 | |
BC021846.1 | 20 | 1986 | AAH21846.1 |
Target Gene Details
Entrez Gene ID: | 18145 |
Gene Name: | Niemann-Pick type C1 |
Gene Aliases: |
A430089E03Rik, C85354, D18Ertd139e, D18Ertd723e, lcsd, nmf164, spm |
Location: |
Chr.18:12189694-12236386 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Npc1 | NM_008720.2 | 25 | 5060 | NP_032746.2 |
AF003348.1 | AAB63372.1 | |||
AK003935.1 | 1 | 921 |