Assay Details
Target Gene Details
Entrez Gene ID: | 224691 |
Gene Name: | zinc finger protein 472 |
Gene Aliases: |
KRIM-1, Krim-1A, Krim-1B, Krim1 |
Location: |
Chr.17:32965831-32979211 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp472 | NM_153063.3 | NP_694703.2 | ||
AK142395.1 | BAE25054.1 | |||
AY195875.1 | AAP34297.1 | |||
BB652931.1 | ||||
BC027407.1 | AAH27407.1 |