Assay Details
Target Gene Details
Entrez Gene ID: | 68816 |
Gene Name: | peptidylprolyl isomerase (cyclophilin)-like 1 |
Gene Aliases: |
1110060O10Rik, AI327391, Cypl1 |
Location: |
Chr.17:29250835-29263971 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppil1 | NM_026845.4 | NP_081121.1 | ||
AK004331.1 | BAB23265.1 | |||
AK150643.1 | BAE29731.1 | |||
AK151572.1 | BAE30513.1 | |||
AK156809.1 | BAE33862.1 | |||
AK189040.1 | ||||
AK217140.1 | ||||
BC058369.1 | AAH58369.1 | |||
BC147647.1 | ||||
BC147649.1 |
More Information
Set Membership: |
Intragenic Non-exonic |