Assay Details
Target Gene Details
Entrez Gene ID: | 27632 |
Gene Name: | negative elongation factor complex member E, Rdbp |
Gene Aliases: |
D17H6S45, NELF-E, RD, Rdbp |
Location: |
Chr.17:34850391-34856372 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nelfe | NM_001045863.1 | NP_001039328.1 | ||
NM_001045864.1 | NP_001039329.1 | |||
NM_138580.2 | NP_613046.2 | |||
AI325150.1 | ||||
AK011663.1 | ||||
AK012389.1 | BAB28206.1 | |||
AK131947.1 | BAE20893.1 | |||
AK147011.1 | BAE27605.1 | |||
BC034867.1 | AAH34867.1 | |||
DV662511.1 |