Assay Details
Target Gene Details
Entrez Gene ID: | 74116 |
Gene Name: | peptidase inhibitor 16 |
Gene Aliases: |
1200009H11Rik, Cripi, PI-16 |
Location: |
Chr.17:29318796-29328903 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pi16 | NM_023734.3 | 2 | 659 | NP_076223.3 |
AK004672.1 | 2 | 353 | ||
AK157854.1 | 2 | 665 | ||
AK158339.1 | 2 | 661 | ||
BC039124.1 | 2 | 256 | AAH39124.1 | |
BC118026.1 | 2 | 307 |
More Information
Set Membership: |
Intragenic Exonic |