Assay Details
Target Gene Details
Entrez Gene ID: | 224480 |
Gene Name: | NADPH oxidase 3 |
Gene Aliases: |
GP91-3, het, nmf250 |
Location: |
Chr.17:3635240-3696261 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nox3 | NM_198958.2 | NP_945196.2 | ||
AY182377.1 | AAO65981.1 | |||
AY573240.1 | AAT80344.1 | |||
BC106862.2 | AAI06863.1 |
More Information
Set Membership: |
Intragenic Non-exonic |