Assay Details
Target Gene Details
Entrez Gene ID: | 116871 |
Gene Name: | metastasis associated 3 |
Gene Aliases: |
1110002J22Rik, mKIAA1266 |
Location: |
Chr.17:83706163-83823715 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mta3 | NM_001171052.1 | 9 | 891 | NP_001164523.1 |
NM_001171053.1 | 9 | 891 | NP_001164524.1 | |
NM_001171054.1 | 9 | 891 | NP_001164525.1 | |
NM_054082.2 | 9 | 888 | NP_473423.1 | |
AF288138.1 | 9 | 842 | AAK83045.1 | |
AK129318.1 | 9 | 820 | BAC98128.1 | |
AK145942.1 | 9 | 836 | BAE26772.1 | |
AK146721.1 | 9 | 844 | BAE27384.1 | |
AK146905.1 | 9 | 844 | BAE27518.1 | |
AK147197.1 | 9 | 834 | BAE27755.1 | |
AK154856.1 | 9 | 831 | BAE32881.1 | |
AK158866.1 | 8 | 754 | BAE34701.1 | |
BC022124.1 | 9 | 855 | AAH22124.1 | |
BC099960.1 | 9 | 808 | ||
CA751287.1 | 3 | 231 |
More Information
Set Membership: |
Intragenic Exonic |